Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165997.1 | NP_001159469.1 | 4 - 5 | 667 | 123 | |
NM_001165998.1 | NP_001159470.1 | 3 - 4 | 531 | 123 | ||
NM_029773.2 | NP_084049.2 | 4 - 5 | 667 | 123 | ||
GenBank mRNA | BC111867.1 | - | 4 - 5 | 541 | 40056321 | 123 |
CF746115.1 | - | 1 - 2 | 120 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |