Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123386.1 | NP_001116858.1 | 2 - 3 | 1154 | 63 | |
NM_009881.3 | NP_034011.1 | 5 - 6 | 1104 | 63 | ||
GenBank mRNA | AF081260.1 | - | 5 - 6 | 1086 | 63 | |
AF081261.1 | - | 5 - 6 | 1086 | 63 | ||
AK028162.1 | - | 2 - 3 | 850 | 63 | ||
AK156509.1 | - | 2 - 3 | 1155 | 63 | ||
BC055103.1 | - | 5 - 6 | 1053 | 6741468 | 63 | |
BC062123.1 | - | 2 - 3 | 1243 | 6826210 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |