Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178358.3 | NP_848135.1 | 1 - 2 | 105 | 139 | |
GenBank mRNA | AK142526.1 | - | 1 - 2 | 162 | 139 | |
AY253666.1 | - | 1 - 2 | 161 | 139 | ||
BC051434.1 | - | 1 - 2 | 105 | 6389932 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |