Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198958.2 | NP_945196.2 | 10 - 11 | 1368 | 94 | |
GenBank mRNA | AY182377.1 | - | 10 - 11 | 1368 | 94 | |
AY573240.1 | - | 10 - 11 | 1308 | 94 | ||
BC106862.2 | - | 10 - 11 | 1368 | 40047310 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |