Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081173.1 | NP_001074642.1 | 6 - 7 | 1053 | 99 | |
GenBank mRNA | AK144916.1 | - | 6 - 7 | 883 | 99 | |
BC056180.1 | - | 1 - 2 | 73 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |