Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290424.1 | NP_001277353.1 | 12 - 13 | 1456 | 141 | |
NM_008031.3 | NP_032057.2 | 12 - 13 | 1456 | 141 | ||
GenBank mRNA | AF461114.1 | - | 12 - 13 | 1180 | 141 | |
AK140343.1 | - | 12 - 13 | 1418 | 141 | ||
BC079671.1 | - | 12 - 13 | 1442 | 30532682 | 141 | |
L23971.1 | - | 12 - 13 | 1303 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |