Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033167.3 | NP_001028339.1 | 7 - 8 | 1909 | 63 | |
GenBank mRNA | AK147397.1 | - | 7 - 8 | 1910 | 63 | |
BC039219.1 | - | 4 - 5 | 478 | 63 | ||
BC041671.1 | - | 4 - 5 | 478 | 63 | ||
BC094372.1 | - | 673 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |