Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190817.1 | NP_001177746.1 | 7 - 8 | 1135 | 94 | |
NM_007869.3 | NP_031895.1 | 7 - 8 | 1135 | 94 | ||
GenBank mRNA | AK133582.1 | - | 7 - 8 | 1117 | 94 | |
AK144228.1 | - | 7 - 8 | 1091 | 94 | ||
BC080300.1 | - | 7 - 8 | 1116 | 94 | ||
L16953.1 | - | 7 - 8 | 812 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |