Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146047.2 | NP_666159.2 | 8 - 9 | 1220 | 131 | |
GenBank mRNA | AK048357.1 | - | 8 - 9 | 1117 | 131 | |
AK156203.1 | - | 8 - 9 | 1111 | 131 | ||
AK165939.1 | - | 8 - 9 | 1081 | 131 | ||
BC026562.1 | - | 8 - 9 | 921 | 5028264 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |