Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ushbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181418.3 | NP_852083.2 | 3 - 4 | 339 | 65 | |
GenBank mRNA | AK034939.1 | - | 2 - 3 | 509 | 65 | |
AK043798.1 | - | 3 - 4 | 329 | 65 | ||
BC026453.1 | - | 3 - 4 | 323 | 4460920 | 65 | |
BY241807.1 | - | 3 - 4 | 355 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |