Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172756.2 | NP_766344.2 | 7 - 8 | 1329 | 137 | |
GenBank mRNA | AK049395.1 | - | 5 - 6 | 1636 | 137 | |
AK078830.1 | - | 6 - 7 | 1559 | 137 | ||
AK080768.1 | - | 5 - 6 | 1642 | 137 | ||
AK083552.1 | - | 5 - 6 | 1628 | 137 | ||
AK087444.1 | - | 5 - 6 | 1647 | 137 | ||
BC030436.1 | - | 7 - 8 | 1268 | 137 | ||
BC094316.1 | - | 7 - 8 | 1286 | 137 | ||
BC145659.1 | - | 3 - 4 | 552 | 137 | ||
EU184014.1 | - | 7 - 8 | 1305 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |