Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167981.1 | NP_001161453.1 | 3 - 4 | 830 | 109 | |
NM_144731.4 | NP_653332.3 | 3 - 4 | 830 | 109 | ||
GenBank mRNA | AF349573.1 | - | 3 - 4 | 754 | 109 | |
AK030683.1 | - | 3 - 4 | 778 | 109 | ||
AK033427.1 | - | 3 - 4 | 779 | 109 | ||
AK036523.1 | - | 3 - 4 | 779 | 109 | ||
AK041791.1 | - | 3 - 4 | 807 | 109 | ||
AK151424.1 | - | 2 - 3 | 421 | 109 | ||
AK151753.1 | - | 2 - 3 | 421 | 109 | ||
AK153143.1 | - | 2 - 3 | 421 | 109 | ||
BC052461.1 | - | 3 - 4 | 806 | 6834581 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |