Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012056.2 | NP_036186.2 | 8 - 9 | 1517 | 79 | |
GenBank mRNA | AF090334.1 | - | 8 - 9 | 1443 | 79 | |
AF279263.1 | - | 8 - 9 | 1495 | 79 | ||
AK007499.1 | - | 6 - 7 | 907 | 79 | ||
AK218657.1 | - | 1 - 2 | 126 | 79 | ||
BC026133.1 | - | 8 - 9 | 1457 | 4018235 | 79 | |
BC043129.1 | - | 8 - 9 | 1517 | 6810029 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |