Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105197.1 | NP_001098667.1 | 5 - 6 | 854 | 113 | |
NM_001271489.1 | NP_001258418.1 | 5 - 6 | 1149 | 113 | ||
NM_001271490.1 | NP_001258419.1 | 5 - 6 | 1321 | 113 | ||
NM_172472.3 | NP_766060.2 | 5 - 6 | 1168 | 113 | ||
GenBank mRNA | AK144657.1 | - | 5 - 6 | 1133 | 113 | |
AK144751.1 | - | 5 - 6 | 1136 | 113 | ||
AK145029.1 | - | 5 - 6 | 114 | |||
AK145832.1 | - | 5 - 6 | 1133 | 113 | ||
AK147405.1 | - | 4 - 5 | 1623 | 113 | ||
AK150372.1 | - | 5 - 6 | 920 | 113 | ||
AK151447.1 | - | 5 - 6 | 839 | 113 | ||
AK152221.1 | - | 5 - 6 | 839 | 113 | ||
AK153051.1 | - | 5 - 6 | 920 | 113 | ||
AK154690.1 | - | 4 - 5 | 1719 | 113 | ||
AK159162.1 | - | 5 - 6 | 1049 | 113 | ||
AK220116.1 | - | 2 - 3 | 149 | 113 | ||
BC056358.1 | - | 5 - 6 | 1156 | 6813486 | 113 | |
BC063047.1 | - | 5 - 6 | 1156 | 6813486 | 113 | |
S76673.1 | - | 3 - 4 | 506 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |