Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024136.1 | NP_001019307.1 | 4 - 5 | 1113 | 70 | |
GenBank mRNA | BC060663.1 | - | 4 - 5 | 1113 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |