Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024136.1 | NP_001019307.1 | 9 - 10 | 1771 | 74 | |
GenBank mRNA | BC060663.1 | - | 10 - 11 | 1975 | 74 | |
BC072562.1 | - | 3 - 4 | 362 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |