Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026917.5 | NP_081193.2 | 5 - 6 | 848 | 77 | |
GenBank mRNA | AB088044.1 | - | 4 - 5 | 614 | 77 | |
AK007355.1 | - | 6 - 7 | 1320 | 77 | ||
AK018755.1 | - | 5 - 6 | 814 | 77 | ||
AK080541.1 | - | 5 - 6 | 814 | 77 | ||
AK169197.1 | - | 795 | 77 | |||
AY166672.1 | - | 4 - 5 | 614 | 77 | ||
BC005531.1 | - | 6 - 7 | 875 | 77 | ||
BC025126.1 | - | 5 - 6 | 833 | 3669723 | 77 | |
BC052464.1 | - | 5 - 6 | 753 | 6834763 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |