Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spaca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085393.2 | NP_001078862.1 | 1 - 2 | 146 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |