Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175114.3 | NP_780323.2 | 11 - 12 | 1331 | 73 | |
GenBank mRNA | AK029123.1 | - | 12 - 13 | 1729 | 73 | |
AK041436.1 | - | 12 - 13 | 1699 | 73 | ||
AK122426.1 | - | 12 - 13 | 1421 | 73 | ||
BC058971.1 | - | 11 - 12 | 1273 | 6832507 | 73 | |
BC116262.1 | - | 11 - 12 | 1298 | 73 | ||
BC116263.1 | - | 11 - 12 | 1298 | 73 | ||
BC129961.1 | - | 11 - 12 | 1298 | 73 | ||
BC129962.1 | - | 11 - 12 | 1298 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |