Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK142152.1 | - | 9 - 10 | 1099 | 90 | |
AK154645.1 | - | 6 - 7 | 3339 | 90 | ||
AK163238.1 | - | 7 - 8 | 2687 | 90 | ||
BC031484.1 | - | 9 - 10 | 1097 | 2654265 | 90 | |
BC044769.1 | - | 2 - 3 | 1042 | 90 | ||
BC046438.1 | - | 1 - 2 | 73 | 90 | ||
BC047372.1 | - | 9 - 10 | 1080 | 90 | ||
BC057597.1 | - | 7 - 8 | 2665 | 90 | ||
BC089379.1 | - | 9 - 10 | 1054 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |