Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007596.2 | NP_031622.2 | 1 - 2 | 252 | 73 | |
GenBank mRNA | AK002895.1 | - | 1 - 2 | 251 | 73 | |
BC005688.1 | - | 1 - 2 | 175 | 3156307 | 73 | |
CF915604.1 | - | 1 - 2 | 273 | 73 | ||
U21960.1 | - | 1 - 2 | 200 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |