Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mefv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161790.1 | NP_001155262.1 | 5 - 6 | 1934 | 61 | |
NM_001161791.1 | NP_001155263.1 | 5 - 6 | 1844 | 61 | ||
NM_019453.2 | NP_062326.2 | 5 - 6 | 1844 | 61 | ||
GenBank mRNA | AF143409.1 | - | 5 - 6 | 1817 | 61 | |
BC108993.1 | - | 5 - 6 | 1805 | 40047838 | 61 | |
BC108994.1 | - | 5 - 6 | 1895 | 40047839 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |