Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Litaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019980.2 | NP_064364.1 | 1 - 2 | 83 | 68 | |
GenBank mRNA | AK018578.1 | - | 1 - 2 | 66 | 68 | |
AK076162.1 | - | 1 - 2 | 85 | 68 | ||
AK078708.1 | - | 1 - 2 | 83 | 68 | ||
AK151053.1 | - | 1 - 2 | 83 | 68 | ||
AK159533.1 | - | 1 - 2 | 51 | 68 | ||
BC018559.1 | - | 1 - 2 | 63 | 2812674 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |