Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285472.1 | NP_001272401.1 | 1 - 2 | 47 | 100 | |
GenBank mRNA | AF349658.1 | - | 1 - 2 | 47 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |