Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172824.3 | NP_766412.2 | 7 - 8 | 840 | 90 | |
GenBank mRNA | AK090294.1 | - | 7 - 8 | 747 | 90 | |
BC031204.1 | - | 7 - 8 | 733 | 5066612 | 90 | |
BC095999.1 | - | 7 - 8 | 736 | 90 | ||
CF742918.1 | - | 5 - 6 | 573 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |