Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010816.1 | NP_034946.1 | 19 - 20 | 2121 | 89 | |
GenBank mRNA | AF084945.1 | - | 19 - 20 | 2121 | 89 | |
BC030893.1 | - | 19 - 20 | 2070 | 4487929 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |