Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144550.4 | NP_653133.3 | 8 - 9 | 866 | 65 | |
GenBank mRNA | AK048789.1 | - | 8 - 9 | 810 | 65 | |
AK165031.1 | - | 6 - 7 | 709 | 65 | ||
AY099108.1 | - | 8 - 9 | 1146 | 65 | ||
BC052381.1 | - | 8 - 9 | 820 | 5703283 | 65 | |
BC056960.1 | - | 8 - 9 | 820 | 5703283 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |