Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026666.3 | NP_080942.1 | 10 - 11 | 2182 | 75 | |
GenBank mRNA | AK147571.1 | - | 10 - 11 | 2183 | 75 | |
BC060723.1 | - | 8 - 9 | 1110 | 75 | ||
BC098372.1 | - | 8 - 9 | 1342 | 30363389 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |