Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
NM_001198823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
NM_001198823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
NM_001198824.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
NM_001198824.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
NM_001198825.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
NM_001198825.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
NM_001198826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
NM_001198826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
NM_007471.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
NM_007471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015248.mm.1 |
TC1600001966.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
NM_001198823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
NM_001198823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
NM_001198824.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
NM_001198824.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
NM_001198825.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
NM_001198825.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
NM_001198826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
NM_001198826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
NM_007471.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
NM_007471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015249.mm.1 |
TC1600001966.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
NM_001198823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
NM_001198823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
NM_001198824.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
NM_001198824.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
NM_001198825.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
NM_001198825.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
NM_001198826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
NM_001198826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
NM_007471.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
NM_007471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600015253.mm.1 |
TC1600001966.mm.1 |
MTA TC |