Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039521.1 | NP_001034610.1 | 16 - 17 | 1759 | 80 | |
GenBank mRNA | AK080680.1 | - | 1740 | 80 | ||
AK165676.1 | - | 16 - 17 | 1747 | 80 | ||
AK170340.1 | - | 1 - 2 | 74 | 80 | ||
BC034110.1 | - | 12 - 13 | 1274 | 80 | ||
BC055781.1 | - | 3 - 4 | 377 | 80 | ||
BC138774.1 | - | 16 - 17 | 1724 | 8861796 | 80 | |
BC138776.1 | - | 16 - 17 | 1724 | 8861798 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |