Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113208.1 | NP_001106679.1 | 12 - 13 | 1871 | 80 | |
NM_010954.4 | NP_035084.1 | 12 - 13 | 1871 | 80 | ||
GenBank mRNA | AF001286.1 | - | 12 - 13 | 1797 | 80 | |
AF001287.1 | - | 12 - 13 | 1797 | 80 | ||
AF016619.1 | - | 12 - 13 | 1753 | 80 | ||
BC119027.1 | - | 12 - 13 | 1729 | 8733776 | 80 | |
BC119029.1 | - | 12 - 13 | 1729 | 8733778 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |