Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013803.2 | NP_038831.2 | 2 - 3 | 703 | 84 | |
GenBank mRNA | AF110178.1 | - | 2 - 3 | 704 | 84 | |
AF110179.1 | - | 2 - 3 | 704 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Probe spans exons |