Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081401.2 | NP_001074870.1 | 4 - 5 | 1106 | 133 | |
NM_177872.2 | NP_808540.2 | 4 - 5 | 1106 | 133 | ||
GenBank mRNA | BC158091.1 | - | 4 - 5 | 707 | 9088176 | 133 |
BC158105.1 | - | 4 - 5 | 707 | 9088181 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |