Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arvcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272028.1 | NP_001258957.1 | 10 - 11 | 2286 | 62 | |
NM_001272029.1 | NP_001258958.1 | 10 - 11 | 2419 | 62 | ||
NM_001272030.1 | NP_001258959.1 | 9 - 10 | 2268 | 62 | ||
NM_001272031.1 | NP_001258960.1 | 9 - 10 | 2029 | 62 | ||
NM_001272032.1 | NP_001258961.1 | 8 - 9 | 2011 | 62 | ||
NM_033474.3 | NP_258435.2 | 11 - 12 | 2437 | 62 | ||
GenBank mRNA | AF286212.1 | - | 10 - 11 | 2286 | 62 | |
AF286213.1 | - | 9 - 10 | 2268 | 62 | ||
AF286214.1 | - | 9 - 10 | 1938 | 62 | ||
AF286215.1 | - | 8 - 9 | 1920 | 62 | ||
AJ243418.1 | - | 9 - 10 | 2053 | 62 | ||
AK031855.1 | - | 4 - 5 | 394 | 62 | ||
AK044982.1 | - | 8 - 9 | 1959 | 62 | ||
AK051627.1 | - | 9 - 10 | 1985 | 62 | ||
AK084886.1 | - | 10 - 11 | 2407 | 62 | ||
BC051655.1 | - | 5 - 6 | 434 | 62 | ||
BC052393.1 | - | 10 - 11 | 2338 | 62 | ||
BC056980.1 | - | 11 - 12 | 2345 | 6821090 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |