Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285800.1 | NP_001272729.1 | 1 - 2 | 180 | 78 | |
NM_022811.3 | NP_073722.1 | 1 - 2 | 180 | 78 | ||
GenBank mRNA | AK050780.1 | - | 128 | 78 | ||
AK083948.1 | - | 1 - 2 | 77 | |||
AK132556.1 | - | 1 - 2 | 151 | 78 | ||
BC034783.1 | - | 1 - 2 | 127 | 4022372 | 78 | |
BY328290.1 | - | 1 - 2 | 182 | 78 | ||
D14336.1 | - | 1 - 2 | 143 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |