Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007509.3 | NP_031535.2 | 11 - 12 | 1219 | 69 | |
GenBank mRNA | AK146499.1 | - | 11 - 12 | 1184 | 69 | |
AK151200.1 | - | 11 - 12 | 1220 | 69 | ||
AK151322.1 | - | 11 - 12 | 1192 | 69 | ||
AK151586.1 | - | 11 - 12 | 1192 | 69 | ||
AK152718.1 | - | 11 - 12 | 1192 | 69 | ||
AK152766.1 | - | 11 - 12 | 1192 | 69 | ||
AK153341.1 | - | 11 - 12 | 1183 | 69 | ||
AK159133.1 | - | 11 - 12 | 1190 | 69 | ||
AK159153.1 | - | 11 - 12 | 1190 | 69 | ||
AK159586.1 | - | 11 - 12 | 1187 | 69 | ||
AK159701.1 | - | 11 - 12 | 1191 | 69 | ||
AK159986.1 | - | 11 - 12 | 1191 | 69 | ||
AK160080.1 | - | 11 - 12 | 1190 | 69 | ||
AK160854.1 | - | 11 - 12 | 1185 | 69 | ||
AK166669.1 | - | 11 - 12 | 1159 | 69 | ||
AK168852.1 | - | 11 - 12 | 1183 | 69 | ||
AK169155.1 | - | 11 - 12 | 1198 | 69 | ||
AK169270.1 | - | 11 - 12 | 1195 | 69 | ||
AK170110.1 | - | 11 - 12 | 1214 | 69 | ||
AK189307.1 | - | 2 - 3 | 154 | 69 | ||
AK190992.1 | - | 2 - 3 | 154 | 69 | ||
AK196704.1 | - | 2 - 3 | 154 | 69 | ||
AK216659.1 | - | 2 - 3 | 154 | 69 | ||
AK218365.1 | - | 2 - 3 | 154 | 69 | ||
BC012497.1 | - | 11 - 12 | 1184 | 4500843 | 69 | |
BC046302.1 | - | 11 - 12 | 1174 | 6492612 | 69 | |
BC085300.1 | - | 11 - 12 | 1166 | 6305977 | 69 | |
EF437356.1 | - | 1 - 2 | 50 | 69 | ||
Y12634.1 | - | 11 - 12 | 1176 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |