Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC003305.1 | - | 6 - 7 | 1198 | 3584788 | 93 |
BC158040.1 | - | 6 - 7 | 1043 | 9088156 | 93 | |
CT010344.1 | - | 6 - 7 | 1011 | 93 | ||
J03302.1 | - | 6 - 7 | 987 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |