Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY293285.1 | - | 1 - 2 | 114 | 94 | |
BC108963.1 | - | 1 - 2 | 132 | 40046744 | 94 | |
BC108964.1 | - | 1 - 2 | 132 | 40046745 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |