Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252679.1 | NP_001239608.1 | 2 - 3 | 141 | 120 | |
GenBank mRNA | U82379.1 | - | 2 - 3 | 141 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |