Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290792.1 | NP_001277721.1 | 2 - 3 | 226 | 57 | |
NM_001290794.1 | NP_001277723.1 | 2 - 3 | 122 | 57 | ||
NM_001290795.1 | NP_001277724.1 | 2 - 3 | 226 | 57 | ||
NM_172372.2 | NP_758960.1 | 2 - 3 | 222 | 57 | ||
GenBank mRNA | AF229635.1 | - | 2 - 3 | 200 | 57 | |
AK018409.1 | - | 2 - 3 | 222 | 57 | ||
AK145616.1 | - | 2 - 3 | 122 | 57 | ||
AK214427.1 | - | 1 - 2 | 61 | 57 | ||
BC011479.1 | - | 2 - 3 | 193 | 4035468 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |