Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017468.3 | NP_059496.1 | 1 - 2 | 211 | 113 | |
GenBank mRNA | BC115848.1 | - | 1 - 2 | 123 | 40048943 | 113 |
U82698.1 | - | 1 - 2 | 212 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |