Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133985.2 | NP_598746.2 | 7 - 8 | 1055 | 156 | |
NR_045693.1 | 8 - 9 | 1184 | 156 | |||
GenBank mRNA | AK075837.1 | - | 6 - 7 | 563 | 156 | |
AK129291.1 | - | 7 - 8 | 1050 | 156 | ||
BC046453.1 | - | 8 - 9 | 1079 | 156 | ||
BC060645.1 | - | 7 - 8 | 1038 | 6852021 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |