Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029857.3 | NP_084133.1 | 2 - 3 | 499 | 123 | |
GenBank mRNA | AK029092.1 | - | 2 - 3 | 499 | 123 | |
AK054337.1 | - | 2 - 3 | 496 | 123 | ||
AK148086.1 | - | 824 | 123 | |||
AK170619.1 | - | 2 - 3 | 228 | 123 | ||
AK171189.1 | - | 2 - 3 | 245 | 123 | ||
BC013471.1 | - | 1 - 2 | 125 | 4221072 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |