Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029857.3 | NP_084133.1 | 3 - 4 | 645 | 66 | |
GenBank mRNA | AK014571.1 | - | 2 - 3 | 246 | 66 | |
AK029092.1 | - | 3 - 4 | 645 | 66 | ||
AK054337.1 | - | 3 - 4 | 642 | 66 | ||
AK148086.1 | - | 970 | 66 | |||
AK170619.1 | - | 3 - 4 | 374 | 66 | ||
AK171189.1 | - | 3 - 4 | 391 | 66 | ||
BC013471.1 | - | 2 - 3 | 271 | 4221072 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |