Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082531.1 | NP_001076000.1 | 3 - 4 | 225 | 130 | |
NR_002926.3 | 3 - 4 | 245 | 131 | |||
GenBank mRNA | AK131857.1 | - | 3 - 4 | 246 | 131 | |
BC045156.1 | - | 3 - 4 | 225 | 6479080 | 130 | |
U28244.1 | - | 3 - 4 | 232 | 130 | ||
U32359.1 | - | 2 - 3 | 200 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |