Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007711.3 | NP_031737.1 | 9 - 10 | 2125 | 71 | |
NM_173873.1 | NP_776298.1 | 10 - 11 | 2301 | 71 | ||
NM_173874.1 | NP_776299.1 | 10 - 11 | 2301 | 71 | ||
NM_173876.3 | NP_776301.1 | 9 - 10 | 1846 | 71 | ||
GenBank mRNA | AF029347.1 | - | 9 - 10 | 1749 | 71 | |
AK141136.1 | - | 10 - 11 | 2239 | 71 | ||
AK149276.1 | - | 10 - 11 | 1840 | 71 | ||
AK150688.1 | - | 10 - 11 | 2230 | 71 | ||
AK159698.1 | - | 10 - 11 | 2275 | 71 | ||
AK161327.1 | - | 10 - 11 | 2229 | 71 | ||
AK162639.1 | - | 10 - 11 | 2242 | 71 | ||
AK169297.1 | - | 10 - 11 | 2222 | 71 | ||
BC057133.1 | - | 9 - 10 | 1779 | 6833732 | 71 | |
BC057855.1 | - | 9 - 10 | 1941 | 71 | ||
X78874.1 | - | 9 - 10 | 1950 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |