Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133972.2 | NP_598733.2 | 1 - 2 | 312 | 67 | |
GenBank mRNA | AK010815.1 | - | 1 - 2 | 340 | 67 | |
AK081550.1 | - | 1 - 2 | 357 | 67 | ||
BC043070.1 | - | 1 - 2 | 312 | 6403587 | 67 | |
BC048396.1 | - | 1 - 2 | 273 | 67 | ||
BC055779.1 | - | 1 - 2 | 312 | 6403587 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |