Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004062.2 | NP_001004062.1 | 7 - 8 | 686 | 83 | |
GenBank mRNA | AK129173.1 | - | 7 - 8 | 83 | ||
BC056137.1 | - | 7 - 8 | 692 | 83 | ||
BC068139.1 | - | 7 - 8 | 692 | 83 | ||
BC080308.1 | - | 7 - 8 | 686 | 5719562 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |