Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135112.1 | NP_001128584.1 | 10 - 11 | 1402 | 97 | |
GenBank mRNA | AF325535.1 | - | 10 - 11 | 1443 | 97 | |
AK011535.2 | - | 3 - 4 | 340 | 97 | ||
AK031250.1 | - | 10 - 11 | 1359 | 97 | ||
AK167511.1 | - | 10 - 11 | 1364 | 97 | ||
BC003920.1 | - | 10 - 11 | 1335 | 97 | ||
BC027240.1 | - | 10 - 11 | 1364 | 2654866 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |