Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025495.3 | NP_079771.1 | 1 - 2 | 147 | 108 | |
GenBank mRNA | AK006234.1 | - | 1 - 2 | 184 | 107 | |
AK012863.1 | - | 1 - 2 | 151 | 108 | ||
AK014919.1 | - | 1 - 2 | 103 | 108 | ||
AK076577.1 | - | 1 - 2 | 185 | 108 | ||
BC094280.1 | - | 1 - 2 | 201 | 108 | ||
BC147207.1 | - | 1 - 2 | 127 | 9056262 | 108 | |
BC147208.1 | - | 1 - 2 | 127 | 9056260 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |